Specifications
- Description:SEM Preparation Stand
- Environmentally Preferable:
- Cat. No.:100501-372
- Supplier no.:76750
Specifications
About this item
Specimen mounts of different sizes are placed in the outer ring of the holder base. The center hole of the base will hold a bottle of conductive adhesive. One side of the base takes 10 of the 1/8" pin mounts; reverse side takes 10 of 10 to 15 mm diameter mounts.