ContactUsLinkComponent
RC Filter Flow Air Dryer, Electron Microscopy Sciences
RC Filter Flow Air Dryer, Electron Microscopy Sciences
Catalog # 100502-812
Minority or Woman-Owned Business Enterprise
undefined
RC Filter Flow Air Dryer, Electron Microscopy Sciences
Catalog # 100502-812
Supplier Number:  74838
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Flow Air Dryer
  • Cat. No.:
    100502-812
  • Supplier no.:
    74838
Specifications
About this item
This RC filter flow air dryer means 5 minutes to perfect flat, and spotless RC prints or films.
A built in air filter eliminates dust. It is thermostatically controlled for optimal drying temperatures. Accommodates four 11×14" prints. Measures 19½×14×7¼" (49.5×35.5×18.5 cm).