ContactUsLinkComponent
Rapid Print Washer, Electron Microscopy Sciences
Rapid Print Washer, Electron Microscopy Sciences
Catalog # 100502-810
Minority or Woman-Owned Business Enterprise
undefined
Rapid Print Washer, Electron Microscopy Sciences
Catalog # 100502-810
Supplier Number:  74387
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Rapid Print Washer
  • Cat. No.:
    100502-810
  • Supplier no.:
    74387
Specifications
About this item
Water from 21 powerful jets runs constantly to separate and wash both sides of prints quickly and completely. Agitating water engages and dilutes the hypo, carrying it through the washer and out the opposite side quickly. The Rapid Print Washer accommodates 10 to 12 full-sized prints in one loading; all size films up to 11 x 14" (28 x 35.5 cm). Internal measures: 13 x 151/2".