Silver Conductive 503, Electron Microscopy Sciences
Catalog # 100496-460
Supplier: Avantor
Minority or Woman-Owned Business Enterprise
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Silver Conductive Adhesive 503
- Size:15 g
- Environmentally Preferable:
- Cat. No.:100496-460
- Supplier no.:12686-15
Specifications
About this item
A high temperature conductive paint.
- Withstands ambient temperatures of over 500°F (260°C)
- Remains flexible over temperature range of –40°F to 500°F
- Highly conductive
- Good adhesion to substrates
- Dries at room temperature
- Ready to use
- Easy to apply
Silver paint 503 is a flexible, high temperature conductive material designed for a wide variety of uses, and adheres to most substrates.