Paraplast Medium, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
Certificates
About this item
Cuts to four micron thickness without crumbling and cracking .
- Handy pellet form melts rapidly
- Cuts to 4 micron thickness with excellent ribbon continuity - no Crumbling or cracking
- Double filtered so no need for filtration before use
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- CAS No.:8002-74-2
- Storage Temperature:RT
- Density:0.8 g/cm³ (6.676 lbs/gal)
- Boiling Point:>315 °C (>599 °F
Specifications
Frequently Bought Together