ONCYTE® is a three-dimensional microarray comprised of a combination of nitrocellulose polymer and proprietary chemistry that provides increased protein affinity and binding
Nitrocellulose film surfaces are manufactured and quality tested to the most exacting standards. To provide an optimal balance between binding capacity and fluorescence sensitivity, two variants of nitrocellulose coatings available:
The ONCYTE® AVID™ nitrocellulose-coated substrates are for applications that require the highest possible protein binding affinity and capacity. Therefore AVID™ slides are characterized by unparalleled protein binding capacity and affinity, making them ideal for colometric and infrared applications that require extremely high levels of binding.
The ONCYTE® NOVA™ nitrocellulose-coated substrates are recommended for applications that require the greatest fluorescent sensitivity but can accept reduced binding capacity. Therefore NOVA™ slides extremely low background fluorescence and an elegant, clean surface make them the most sensitive slides available for fluorescence – dependent arrays.
It is also provides enhanced fluorescent chemiluminescent, radiographic, colormetric and visual signal compared to glass or other coated glass surfaces. Nitrocellulose-coated (NC) slides for protein microarrays, cell lysate arrays, tissue printing and other applications.