Orders LinkContactUsLinkComponent
Neon Bulb Array, Electron Microscopy Sciences
Neon Bulb Array, Electron Microscopy Sciences
Neon Bulb Array
Catalog # 100495-474
Supplier #:  97036-01
Neon Bulb Array, Electron Microscopy Sciences
Catalog # 100495-474
Neon Bulb Array, Electron Microscopy Sciences
Neon Bulb Array
Price
$224.62
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This Neon Bulb Array is made from silicone
Each bulb in the mat is 2.5 cm apart. The mat is divided into quadrants. A mark on the back left corner of the array lid is used to key the back left corner of the oven. There are letters and numbers along each edge for easy placement on the Alpha-numeric tray. The mat size is 8 x 8".
Specifications
  • Description:
    Neon Bulb Array