MAC X-ray Microanalysis NBS Standards, Electron Microscopy Sciences
About this item
All MAC X-ray Microanalysis standards are suitable for electron-beam energy dispersive or wavelength dispersive X-ray microanalysis systems. All of our standards are polished to a 0.25 micron diamond finish and carbon coated. They are supplied with fully authenticated certificates of analyses and a location map for standard identification.
NBS glass SRM 481
NBS glass
SRM 482 Au/Cu Set of 6 Standards, comprises 4 alloys and 2 pure metals.
NBS Standard SRM 1872
Set of 3 glasses K-453, K491, K968.
2 Options Available Below
Product Details & Documents
All MAC X-ray Microanalysis standards are suitable for electron-beam energy dispersive or wavelength dispersive X-ray microanalysis systems. All of our standards are polished to a 0.25 micron diamond finish and carbon coated. They are supplied with fully authenticated certificates of analyses and a location map for standard identification.
NBS glass SRM 481
NBS glass
SRM 482 Au/Cu Set of 6 Standards, comprises 4 alloys and 2 pure metals.
NBS Standard SRM 1872
Set of 3 glasses K-453, K491, K968.
Product Family Options
Product Information
- AvailabilityPrice
- 100495-238NBS SRM 481Supplier Number: 80076-01
- 100495-240NBS SRM 482Supplier Number: 80076-02
- 100495-242NBS SRM 1872NBS SRM 1872Discontinued-Supplier Number: -