Orders LinkContactUsLinkComponent
Multi-Vials, Electron Microscopy Sciences
Multi-Vials, Electron Microscopy Sciences
Multi-Vial, Yes
Catalog # 100491-994
Supplier #:  72659
Multi-Vials, Electron Microscopy Sciences
Catalog # 100491-994
Multi-Vials, Electron Microscopy Sciences
Multi-Vials, Electron Microscopy Sciences
Multi-Vial, Yes
Price
$33.64
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Multi-vials are excellent multipurpose containers. They are primarily used for liquid scintillation counting, beta-gamma counting, and sample storage.
  • Nominal volume of 5 ml
  • High density white polyethylene snap cap
  • Alternate white plug cap
They are made from a new type of polypropylene, which assures superior visibility. They come with white polyethylene caps and are perfect and convenient for easy marking.
Specifications
  • Cap included:
    Yes
  • Description:
    Multi-Vial
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Snap Cap for Multi-Vials
    Snap Cap for Multi-Vials
    100500-204
    Log in for availability
    Case of 1,000 - $46.38