Orders LinkContactUsLinkComponent
Microwave Leakage Detector, Electron Microscopy Sciences
Microwave Leakage Detector, Electron Microscopy Sciences
Microwave Leakage Detector
Catalog # 100493-612
Supplier #:  72083-00
Microwave Leakage Detector, Electron Microscopy Sciences
Catalog # 100493-612
Microwave Leakage Detector, Electron Microscopy Sciences
Microwave Leakage Detector, Electron Microscopy Sciences
Microwave Leakage Detector
Price
$500.54
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Evaluate the RF emissions of mobile phone, radio and TV transmission towers, wireless camera, Wi-Fi and LAN detection and installation, microwave oven leakage and other environmental safety concerns.
  • Display also indicates overload and low battery
Radio Frequency (RF) is the range of electromagnetic frequencies above the audio range and below infrared light. Features maximum and minimum hold, an audible alarm, and a zero adjustment to eliminate background EMF.
Dimensions: 130 x 56 x 38 mm. Weight: 170 g.
Delivery information: Comes with a soft carrying case and standard 9 V battery, which provides approximately 100 hours of use.
Specifications
  • Description:
    Microwave Leakage Detector