ContactUsLinkComponent
Microscope Slide Dispenser, Electron Microscopy Sciences
Microscope Slide Dispenser, Electron Microscopy Sciences
Catalog # 100491-770
Minority or Woman-Owned Business Enterprise
undefined
Microscope Slide Dispenser, Electron Microscopy Sciences
Catalog # 100491-770
Supplier Number:  71300
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Slide Dispenser
  • Cat. No.:
    100491-770
  • Supplier no.:
    71300
Specifications
About this item
Molded of attractive gray ABA plastic.
This dispenser provides a method of releasing clean, dust free 1 x 3" slides, one at a time. Comes with a clear molded plastic cover to allow for visual checking of unused slides. Holds approximately 72 slides.
Measurements: 4 3/8 x 4 7/8 x 5 1/2".