Orders LinkContactUsLinkComponent
Micro-Mill® Grinder, Electron Microscopy Sciences
Micro-Mill® Grinder, Electron Microscopy Sciences
Certificates
Micro-Mill® Grinder, Electron Microscopy Sciences
Micro-Mill® Grinder, Electron Microscopy Sciences
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Micro-Mill® Grinder can be used for fast, high-speed grinding of small samples from 20 to 50 mm (¾ to 2")
Micro-Mill® 1: With stainless steel blade, 0-5 minutes electrical timer with an ON position for continuous operation.
Micro-Mill® 2: With hard, stellate faced blade, with a momentary hold-down switch.
The unit comes with a stainless steel or hard-faced blade which rotates at speeds up to 12,000 rpm and will successfully grind bauxite, bone, ceramics, coal, Devorda's alloy, fertilizer, filter paper, fly ash, frozen tissue, glass, grain, kaolinites, leaves, limestone, muscle tissue, pharmaceuticals, pigment blends, plastics, rocks, roots, seeds, slags, soil, teeth, wood, etc. The grinding chamber measures diameter x depth: 54 x 25 mm (2 1/8 x 1"). Its overall dimensions are 14 x 18 x 28 cm (5 ½ x 7 x 11"). It comes with a 1.8 m (6 ft.)3-wire cord and grounded plug. Power: 115V, 60Hz or 220V, 60Hz. The 1/5 horsepower (150 watts) motor develops a torque of 30-ounce-inches (2.2 Kg-cm) at 12,000 rpm.
Product Family Options
Product Information
  • Electrical
    Availability
    Price
  • 100496-818
    Grinder 2
    Grinder 2
    115 V
    Discontinued
    Supplier Number: -
  • 100496-814
    Grinder 1
    Grinder 1
    115 V
    Discontinued
    Supplier Number: -
  • 100496-816
    Grinder 1
    Grinder 1
    220 V
    Log in for availability
    Log in for availability
    Each
    Each - $3,128.46
    $3,128.46
    Each
       $3,128.46
    Supplier Number: 50100-02