Order Entry
United States
ContactUsLinkComponent
Micro-Bed Resin, Electron Microscopy Science
Micro-Bed Resin, Electron Microscopy Science
Catalog # 100503-856
Minority or Woman-Owned Business Enterprise
CAS Number:  
Micro-Bed Resin, Electron Microscopy Science
Catalog # 100503-856
Supplier Number:  14210
Minority or Woman-Owned Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Micro-Bed Embedding Resin
  • Size:
    500 ml
  • Environmentally Preferable:
  • UN No.:
    1993
  • Density:
    1.02006 g/cm³ (8.512 lbs/gal)
  • Boiling Point:
    60 °C (140 °F)
  • Cat. No.:
    100503-856
  • Supplier no.:
    14210

Specifications

About this item

Scala C

Micro-Bed resin is a newly developed water-soluble resin which is a mixture of acrylate and polyester resins. It is a very universal embedding medium and may be used for either light or electron microscopy.

Due to its non-crosslinking characteristics, its hydrophilic nature, and roughness of its section surface, our Micro-Bed Resin is excellent in preserving tissue structure, offers good results in immunolabeling, and stands up well to conventional stains.

In light microscopy, tissues were stained with conventional histological stains (e.g.,hematoxylin-eosin) and embedded in Micro-Bed: the results were comparable with those provided on paraffin embedded tissues. Further, polychrome silver-based and cytochemical stains (e.g.,PAS) were also feasible. In transmission electron microscopy, images as good as those obtained with similar acrylic resins, (e.g., Lowicryl K4M), were obtained.

Micro-Bed is supplied as a single component-ready to use pre-mixed resin. Micro-Bed can be polymerized in either EMS embedding capsules or gelatin capsules by either heat(55-65°C) or UV (wavelength 365nm) light at low temperatures (-10 to 20°C) for 72 hours.

Scala C. et al.(1992). Development of a new acrylic resin ideally suited for light and electron microscopy. Electron Microscopy. Volume 3.EUREM 92, Granada, Spain. pg. 271