Orders LinkContactUsLinkComponent
Measure for Measure, Electron Microscopy Sciences
Measure for Measure, Electron Microscopy Sciences
Measure for Measure Book
Catalog # 100501-574
Supplier #:  31200
Measure for Measure, Electron Microscopy Sciences
Catalog # 100501-574
Measure for Measure, Electron Microscopy Sciences
Measure for Measure, Electron Microscopy Sciences
Measure for Measure Book
Price
$36.12
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
At last, a GREAT Conversion Factor Handbook!
Contains over 39,000 conversions for over 5100 different units.
864 pages
Comprehensive SI reference (the International Standard)
Thousands of foreign and ancient conversion factors
Accurate to 8 significant figures
Value listed in both standard and scientific notation
Handy 4 x 6” size with a durable Lexotone cover
Specifications
  • Description:
    Measure for Measure Book