ContactUsLinkComponent
MAG*I*CAL™ Calibration Standard, Electron Microscopy Sciences
MAG*I*CAL™ Calibration Standard, Electron Microscopy Sciences
Catalog # 100492-378
Minority or Woman-Owned Business Enterprise
CAS Number:  
undefined
MAG*I*CAL™ Calibration Standard, Electron Microscopy Sciences
Catalog # 100492-378
Supplier Number:  80069
Minority or Woman-Owned Business Enterprise
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Magnification Calibration Standard
  • Cat. No.:
    100492-378
  • Supplier no.:
    80069
Specifications
About this item
The MAG*I*CAL™ is a magnification calibration standard for transmission electron microscopes.
  • Performs all major TEM calibrations for all magnification ranges
  • Camera constant
  • Image diffraction pattern rotation
  • Directly traceable to a natural constant
MAG*I*CAL® is the world’s smallest ruler and have been inducted into the Guinness Book of World Records.
MAG*I*CAL™ consists of an ion milled cross section of a silicon single crystal consisting of a series of atomically flat layers of Si and SiGe, which have been grown epitaxially by MBE (molecular beam epitaxy). When the calibration structure is viewed in a TEM, it appears as a series of light and dark layers where the layer thicknesses are accurately known. The calibrated thickness measurements of these light (silicon) and dark (SiGe) layers are based on careful TEM measurements of the lattice spacing of silicon, which is visible on the calibration sample itself, and are supported by x-ray diffraction measurements. The layer spacing are designed so that the sample can be used to calibrate the entire magnification range in a TEM, from 1000x to 1000000x. As the sample is also a single crystal of silicon, the calibrations requiring electron diffraction information such as the camera constant and image/diffraction pattern rotation can also be performed easily and unambiguously. One single calibration sample can therefore be used to provide all three of the major TEM instrument calibration at all magnifications and all camera lengths.