About this item
The same as our Lowicryl complete kits but this new formulation allows you one stop embedding without all of the cumbersome preparation and mixing. This pre-mixed, ready to use resin, saves you time and minimizes chemical contact. MonoStep Lowicryl is ideal for use in immunohistochemistry and immunolabeling. Store at room temperature.