Order Entry
United States
ContactUsLinkComponent
Low Viscosity Mounting Media Kit, by Dr. Spurr, Electron Microscopy Sciences
Low Viscosity Mounting Media Kit, by Dr. Spurr, Electron Microscopy Sciences
Catalog # 100503-444
Minority or Woman-Owned Business Enterprise
CAS Number:  
undefined
Low Viscosity Mounting Media Kit, by Dr. Spurr, Electron Microscopy Sciences
Catalog # 100503-444
Supplier Number:  14300
Minority or Woman-Owned Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Low Viscosity Embedding Kit
  • Environmentally Preferable:
  • Cat. No.:
    100503-444
  • Supplier no.:
    14300

Specifications

About this item

Provides excellent penetration for embedding biological tissues and rapid infiltration. Easy to prepare. The hardness is adjusted by changing the proportion of the flexibilizer (DER 736). The blocks have good trimming and sectioning qualities. Thin sections are tough under the electron beam.

Kit consists of: 450 mL NSA, 225 mL ERL 4221 , 225 mL DER 736 , and 25 mL DMAE.
Spurr, A.R., (1969) J. Ultrastruct. Res. 26, 31-42