Order Entry
United States
ContactUsLinkComponent
Long Molybdenum Boat, Electron Microscopy Sciences
Long Molybdenum Boat, Electron Microscopy Sciences
Catalog # 100488-506
Minority or Woman-Owned Business Enterprise
undefined
Long Molybdenum Boat, Electron Microscopy Sciences
Catalog # 100488-506
Supplier Number:  73818
Minority or Woman-Owned Business Enterprise

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Long Molybdenum Boat
  • Cat. No.:
    100488-506
  • Supplier no.:
    73818

Specifications

About this item

Long Molybdenum Boat of 0.05 mm (0.002") thickness and 75 mm long.

  • Trough measures 50 mm (L) x 5 mm (W) x 1 mm (D).