ContactUsLinkComponent
Kodak Stirring Paddle, Electron Microscopy Sciences
Kodak Stirring Paddle, Electron Microscopy Sciences
Catalog # 100492-196
Minority or Woman-Owned Business Enterprise
undefined
Kodak Stirring Paddle, Electron Microscopy Sciences
Catalog # 100492-196
Supplier Number:  74394
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Paterson Chemical Mixer
  • Cat. No.:
    100492-196
  • Supplier no.:
    74394
Specifications
About this item
This ergonomically designed mixer is specially shaped for efficient stirring and incorporates a particle crusher.
It is invaluable when making up solutions either from liquid concentrates or from dry powdered chemicals.