Specifications
- Capacity:16 oz.
- Description:Photo-Flo 200 Solution
- Cat. No.:100496-354
- Supplier no.:74257
Specifications
About this item
Photo-Flo Solution decreases water surface tension and minimizes water marks and drying streaks on film. It speed dries.