ContactUsLinkComponent
Brain Matrix Blades, Electron Microscopy Sciences
Catalog # 100501-894
Minority or Woman-Owned Business Enterprise
undefined
Brain Matrix Blades, Electron Microscopy Sciences
Catalog # 100501-894
Supplier Number:  70933-70
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Brain Matrix Blades
  • Cat. No.:
    100501-894
  • Supplier no.:
    70933-70
Specifications
About this item
These razor blades are specifically made to work and fit all of our Brain Matrices.
Blades measure 118 x 19 x 0.2286 mm (4.65 x 0.75 x 0.0089"). They are packed 25 blades in a box.