Sure Shot® Spray-Safe Atomizer, Electron Microscopy Sciences
Sure Shot® pressure sprayers are available in 16, 8 and 7 oz. capacities.
- Plated aluminum
- Refillable and rechargeable
- Working pressure: 80 to 145 psi
- Measures are the same as aerosol cans (~2³/₄" Ø)
- Liquid spray lighter than #10 motor oil
All sprayers come standard with Viton® seals, these seals are compatible with most products.
Sure Shot® pressure sprayers are available in 16, 8 and 7 oz. capacities.
- Plated aluminum
- Refillable and rechargeable
- Working pressure: 80 to 145 psi
- Measures are the same as aerosol cans (~2³/₄" Ø)
- Liquid spray lighter than #10 motor oil
All sprayers come standard with Viton® seals, these seals are compatible with most products.
Product Family Options
- DescriptionSize
- : 64186-20
- : 64186-22
- Stainless Steel. Atomizer Model 8500-Pl453.6 g (16 oz.)-: -
- : 64189
- : 64187