ContactUsLinkComponent
Ion Mill Stage, Model 5901, Electron Microscopy Sciences
  100496-872
Minority or Woman-Owned Business Enterprise
undefined
Ion Mill Stage, Model 5901, Electron Microscopy Sciences
  100496-872
 :  50118-01
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Ion Mill Stage, Model 5901
  • Cat. No.:
    100496-872
  • Supplier no.:
    50118-01
The Ion Mill Stage is designed to accept the special SEM stub used with the Tripod Polisher®. The SEM stub is mounted to the Ion Mill Stage and the samples are briefly ion milled in order to remove fine scratches, polishing debris and to give the surface topography prior to SEM analysis.