Orders LinkContactUsLinkComponent
Inoculating Needle, Electron Microscopy Sciences
Inoculating Needle, Electron Microscopy Sciences
Inoculating Needle, 70 mm
Catalog # 100502-490
Supplier #:  62432-01
Inoculating Needle, Electron Microscopy Sciences
Catalog # 100502-490
Inoculating Needle, Electron Microscopy Sciences
Inoculating Needle, Electron Microscopy Sciences
Inoculating Needle, 70 mm
Price
$301.91
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from 26 gauge B and S platinum wire with 15% iridium to provide rigidity with thinner wire for delicate work. Both ends are ground smooth.
Specifications
  • Description:
    Inoculating Needle
  • Length:
    70 mm