Order Entry
United States
ContactUsLinkComponent
GMA/PEG, Electron Microscopy Science
GMA/PEG, Electron Microscopy Science
Catalog # 100504-908
Minority or Woman-Owned Business Enterprise
CAS Number:  
GMA/PEG, Electron Microscopy Science
Catalog # 100504-908
Supplier Number:  14250
Minority or Woman-Owned Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    GMA-Peg Embedding Kit
  • Storage Temperature:
    RT
  • Density:
    1.2025 g/cm³ (10.035 lbs/gal)
  • Boiling Point:
    100 °C (212 °F)
  • Cat. No.:
    100504-908
  • Supplier no.:
    14250

Specifications

About this item

Kit consists of: 450 mL Glycol methacrylate, GMA, 225 mL n-Butyl methacrylate, 50 mL Ethylene glycol dimethacrylate, 15g Benzoyl Peroxide Paste, and 20 mL Polyethylene Glycol 400.

Polyethylene Glycol - Glycol methacrylate mixture. A water miscible embedding medium for cytochemical studies and enzyme localization when light and electron microscopy need to be correlated.
Spaur, R.C. and Moriarty, G., (1977) J Histochem. Cytochem., 23:16