ContactUsLinkComponent
Glass Knife Scoring Jig, Electron Microscopy Sciences
Glass Knife Scoring Jig, Electron Microscopy Sciences
Catalog # 100498-010
Minority or Woman-Owned Business Enterprise
undefined
Glass Knife Scoring Jig, Electron Microscopy Sciences
Catalog # 100498-010
Supplier Number:  62192-02
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Glass Knife Scoring Jig Set
  • Cat. No.:
    100498-010
  • Supplier no.:
    62192-02
Specifications
About this item
These set of jigs assist in the production of making glass knives by hand. When the edge of the T-square jig is placed against the edge of the glass knife strip, the  subsequent score will be 26 mm (1") long. After breaking, the square of the glass can be scored at a 45 degree angle by using the other jig.