Orders LinkContactUsLinkComponent
General Purpose Cleaner, Electron Microscopy Sciences
General Purpose Cleaner, Electron Microscopy Sciences
General Purpose Cleaner, Electron Microscopy Sciences
Certificates
General Purpose Cleaner, Electron Microscopy Sciences
General Purpose Cleaner, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A concentrated solution, which is biodegradable, phosphate and caustic-free alkaline cleaner formulated for general purpose and normal maintenance cleaning applications.
General purpose solution removes general soils, fingerprints, dust, packaging particulates, light oils, and greases. GP solution is safe for use with most base metals and alloys, thermoset and thermoplastic resins, and other synthetic materials. GP solution is easily mixed with water to form a concentration of 10 to 12% by volume and can be used at temperatures ranging from ambient to 82 °C (180 °F).
Product Family Options
Product Information
  • Size
    Packaging
    Availability
    Price
  • 100488-216
    General Purpose Cleaner
    General Purpose Cleaner
    0.25 gal.
    Bottle
    Log in for availability
    Log in for availability
    Each
    Each - $43.88
    $43.88
    Each
       $43.88
    Supplier Number: 71050
  • 100491-400
    General Purpose Cleaner
    General Purpose Cleaner
    1 gal.
    Bottle
    Log in for availability
    Log in for availability
    Gallon
    Gallon - $87.09
    $87.09
    Gallon
       $87.09
    Supplier Number: 71051