About this item
The dryer is designed to rapidly dry slides by blowing warm air over the surface of the slides.
- Temperature control dial allows the user to set the temperature from ambient to 75°C
- Hinged lid made from durable stainless steel
- Zinc coated steel casing with a stoved epoxy finish
- Glass front for see-through viewing
- Self-resetting thermal switch to protect the unit from electrical surges
The slide dryer accommodates two slide racks directly from a stainer and allows the slides to be stained in approximately 20-30 minutes.