About this item
This cloth has been especially designed for long life.
- It is recommended for all final polishing
Made from a rayon flock on a cotton canvas backing. Available with Pressure Sensitive Adhesive (PSA) with and without hole.