Specifications
- Description:Evaporated Pt/Ir on Holey Carbon
- Cat. No.:100496-366
- Supplier no.:80020
Specifications
About this item
Evaporated Pt/Ir on Holey carbon film.
The dots of evaporated Pt/Ir provide dense particles for resolution checks through the particle seperation test.
Holey carbon film support provides holes for ease of focus and astigmatism correction.