ContactUsLinkComponent
EMS Diamond Knife Wash, Electron Microscopy Sciences
EMS Diamond Knife Wash, Electron Microscopy Sciences
  100491-636
Minority or Woman-Owned Business Enterprise
undefined
EMS Diamond Knife Wash, Electron Microscopy Sciences
  100491-636
 :  70605
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Diamond Knife Wash
  • Cat. No.:
    100491-636
  • Supplier no.:
    70605
This new concept of washing the Diamond knives using magnetic stirring action (with special stirring bar) gives you a fast, clean and safe method to remove particles that are left on the knives edge after sectioning.
Since most of the EM labs already have stirring plates, the Diamond Knife Wash has been designed for use with your existing unit, saving you the need to purchase an extra one. It is universal and can be used for all type and brands of Diamond Knives, including the Diatome Histo Knife.
Warning: Never use on a hot plate. If used on a combination Stirrer/Hot Plate, make sure the heat is off.