Specifications
- Description:DER 332/732 Kit
- Cat. No.:100503-428
- Supplier no.:14000
Specifications
About this item
Three different mixtures: one for relatively soft blocks, another for harder blocks for collagenous tissue, and a tougher one if required., Lockwood, W.R. Anatomical Record, 150, 129 (1964). Kit consists of 2x225 ml DER 332, 225 ml DER 732, 450 ml DDSA; Specially Distilled, 50 ml DMP-30.