ContactUsLinkComponent
Combined TEM Test Specimen, Electron Microscopy Sciences
Combined TEM Test Specimen, Electron Microscopy Sciences
Catalog # 100492-364
Minority or Woman-Owned Business Enterprise
undefined
Combined TEM Test Specimen, Electron Microscopy Sciences
Catalog # 100492-364
Supplier Number:  80043
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Combined Test Specimen
  • Cat. No.:
    100492-364
  • Supplier no.:
    80043
Specifications
About this item
This transmission electron microscopy can be used to evaluated microscope performance and contamination.
  • Particles viewed over the holes
  • Used for assessment of factors limiting the microscope performance
A holey carbon film is shadowed with gold and graphitized carbon particles are deposited on a 3.05 mm grid. The evaporated gold forms small polycrystalline islands and within these islands lattice fringes can be resolved. Also use this specimen for the measurement of contamination rates in the EM by noting the deposition rate of carbon within the holes found in the gold film.