To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Coated Lead Shield
- Cat. No.:100496-224
- Supplier no.:64460
Specifications
About this item
This ¼" (6.4 mm) thick lead shield protects against chemical reactions and nuclear radiation.
- W×D×H: 8½×3×6" (22×8×15 cm)
The shield is coated with vinyl for a soft smooth surface, which eliminates the hazard of any skin contact as well as flying debris.