Orders LinkContactUsLinkComponent
Cleaning Basket, Electron Microscopy Sciences
Cleaning Basket, Electron Microscopy Sciences
Cleaning Basket
Catalog # 100496-146
Supplier #:  62110
Cleaning Basket, Electron Microscopy Sciences
Catalog # 100496-146
Cleaning Basket, Electron Microscopy Sciences
Cleaning Basket, Electron Microscopy Sciences
Cleaning Basket
Price
$11.83
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from stainless steel. Keeps small parts together when cleaning ultrasonically. A spring handle applies firm pressure to the basket to keep it closed when in use. Its overall length is 6". Spherical basket has a 38 mm (1 1/2") diameter.
Specifications
  • Description:
    Cleaning Basket