About this item
Vertical Cabinet: Made from clear methyl-methacrylate (Acrylic), this auto-desiccator cabinet features a permanent desiccant contained in the control unit; therefore, no desiccant tray is needed
Non-Vacuum with Dial Hygrometer in Door, Electrically Operated
Vertical Cabinet: Made from clear methyl-methacrylate (Acrylic), this auto-desiccator cabinet features a permanent desiccant contained in the control unit; therefore, no desiccant tray is needed. The cycle is indicated by a green light for a 4 1/2 hours absorption period, or a red light indicating 1/2 hour regeneration of the desiccant. This cabinet will lower relative humidity to 30 - 40% and it includes two adjustable shelves. The unit draws 6 watts, 120 VAC. The Unit measures overall: 31.75 x 39.37 x 61.6 cm (12 1/2 x 15 1/2 x 24 1/2”).
Horizontal Cabinet: The same construction and all features as the Vertical Cabinet but its layout is horizontal. The generator unit is on the side. The overall unit measures: 61.6 x 39.37 x 32.93 cm (24 1/2 x 15 1/2 x 13 1/2”).
Horizontal Cabinet: The same construction and all features as the Vertical Cabinet but its layout is horizontal. The generator unit is on the side. The overall unit measures: 61.6 x 39.37 x 32.93 cm (24 1/2 x 15 1/2 x 13 1/2”).
The cycle is indicated by a green light for a 4 1/2 hours absorption period, or a red light indicating 1/2 hour regeneration of the desiccant. This cabinet will lower relative humidity to 30 - 40% and it includes two adjustable shelves. The unit draws 6 watts, 120 VAC. The Unit measures overall: 31.75 x 39.37 x 61.6 cm (12 1/2 x 15 1/2 x 24 1/2”).