ContactUsLinkComponent
Apiezon Grease; Type M, Electron Microscopy Sciences
Apiezon Grease; Type M, Electron Microscopy Sciences
Catalog # 100504-350
Minority or Woman-Owned Business Enterprise
undefined
Apiezon Grease; Type M, Electron Microscopy Sciences
Catalog # 100504-350
Supplier Number:  60700
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Grease Type M
  • Size:
    25 g
  • Cat. No.:
    100504-350
  • Supplier no.:
    60700
Specifications
About this item
Similar to grease type L, but it contains more wax. Grease M has an estimated vapor pressure at 20°C of 2x10-9 torr, higher than Type L. Type M is recommended for applications where a grease of moderately low vapor pressure is required; such as for sealing ground joints on the backing side of oil diffusion pumps.