ContactUsLinkComponent
Apiezon Grease; Type L, Electron Microscopy Sciences
Apiezon Grease; Type L, Electron Microscopy Sciences
Catalog # 100504-354
Minority or Woman-Owned Business Enterprise
undefined
Apiezon Grease; Type L, Electron Microscopy Sciences
Catalog # 100504-354
Supplier Number:  60702
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Grease Type L
  • Size:
    25 g
  • Cat. No.:
    100504-354
  • Supplier no.:
    60702
Specifications
About this item
Grease L is a petroleum hydrocarbon grease, containing no additives. Very low vapor pressure (at 20°C of 8x10-11 torr), high vacuum grade. Type L is widely used in the stationary phase of gas liquid chromatography, and may be used on all ground joints in a vacuum system (not for stopcocks). Its solubility is as follows: in Ethanol at 20°C-0.003%; 60°C-0.005%; 100°C-0.011%; 150°C-0.011%. Grease L softens and melts at about 47°C and it is not recommended for joints where temperatures exceed more than 30°C in use. Grease T is recommended for these higher temperatures.