Aperture Flamers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
For holding 1/4 to 1/2" Ø aperture discs while flaming.
- Small discs can be flamed in the 3/16" Ø cup made from 6 mil platinum foil with a 1/32" Ø hole in the bottom to induce a good flow of air
- Cup is attached by a 30 mil platinum wire to a heat resistant handle
The aperture can be hung from the hook for flaming.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.