About this item
An Anti-Roll (A/R) Device which may be used with all our blade holders.
It snaps on or off by a unique magnetic attachment system. Its clear plastic plate which is positioned above the sectioning blade prevents any sample curling.