ContactUsLinkComponent
Anemometer, Electron Microscopy Sciences
Anemometer, Electron Microscopy Sciences
Catalog # 100501-344
Minority or Woman-Owned Business Enterprise
undefined
Anemometer, Electron Microscopy Sciences
Catalog # 100501-344
Supplier Number:  72082
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Anemometer
  • Cat. No.:
    100501-344
  • Supplier no.:
    72082
Specifications
About this item
Perfect for use in fume hoods, environmental studies, or for air flow check. A highly accurate, hand-held unit, only 3 ozs. Allows you to select readout in knots, feet per minute, meters per second, or miles per hour. To insure maximum sensitivity and accuracy, the turbine is suspended on saphire jewel bearings, and its rotation is sensed by an infrared light beam, which adds no friction. The resulting signal is pro-cessed electrically by an LSI (Large Scale Integrated) circuit. Measurements: 4.5 x 2.5 x 1.5".