ContactUsLinkComponent
Alpha-Numeric Oven Tray, Electron Microscopy Sciences
Alpha-Numeric Oven Tray, Electron Microscopy Sciences
Catalog # 100495-472
Minority or Woman-Owned Business Enterprise
Alpha-Numeric Oven Tray, Electron Microscopy Sciences
Catalog # 100495-472
Supplier Number:  97035-01
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Alpha-Numeric Oven Tray
  • Cat. No.:
    100495-472
  • Supplier no.:
    97035-01
Specifications
About this item
Made from polypropylene, each line is spaced 1 cm apart with letters along the sides and numbers across the top and bottom. This tray has ruled lines that aid in the reproducible placement of the specimens in the oven.
Developed in collaboration with Dr. Gary Login, Beth Israel Hospital, Boston, MA.