About this item
The original anionic powder detergents for manual and ultrasonic cleaning. Odorless, high sequestering powder. These helps to decontaminate radioactive surfaces.
- Cost efficient
- Versatile
Alconox detergents safely cleans glass, plastic, metal, rubber, and porcelain without leaving a film, residue, or any scratches.