3-Well Porcelain Micro Spot Plate, Electron Microscopy Sciences
Porcelain Micro Spot Plate is ideal for microchemical applications. The plate has three concave depressions.
- Cavities measure O.Ø×Depth 22×7 mm
- Plate overall measures 85×34 mm
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.