About this item
Long shanks, strong tips. Length: 180 mm
Antimagnetic is composed of C, Cr, Ni and Mo. It offers a good resistance to corrosion thanks to its high concentration of molybdenum and is 80% antimagnetic but is not as hard as Inox. Antimagnetic is resistant up to temperatures of around 400°C (DIN 50 914) and is suitable for autoclave sterilization at 270°C.
Polished Inox 2 is a standard magnetic stanless steel that is not as hard as carbon steels because it contains chromium. Can not be sterilized.