Orders LinkContactUsLinkComponent
Quick-Test Auto-Scanning Tweezers, Electron Microscopy Sciences
Quick-Test Auto-Scanning Tweezers, Electron Microscopy Sciences
Quick-Test Auto-Scanning Tweezers, Straight Tips
Catalog # 102098-076
Supplier #:  72907-01
Quick-Test Auto-Scanning Tweezers, Electron Microscopy Sciences
Catalog # 102098-076
Quick-Test Auto-Scanning Tweezers, Electron Microscopy Sciences
Quick-Test Auto-Scanning Tweezers, Electron Microscopy Sciences
Quick-Test Auto-Scanning Tweezers, Straight Tips
Price
$108.10
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Smart tweezers is a LCR meter in a set of tweezers. The Smart Tweezers feature a unique, patented mechanical and electronic design that integrates a highly accurate digital multimeter with built-in high precision SMD probes and a display.
  • 10 times lighter than conventional tools
  • Flexible user interface that allows the user to configure default settings
  • Measures DC voltage up to 8 volts
  • Measures AC voltage via an oscilloscope-like picture
  • Measures inductance, and resistance
  • Ergo design convenient control
  • Tips are made from gold-plated, non-magnetic stainless steel
Continuity/Open testing-beeper sounds when a resistance reading is below a threshold or to indicate an open circuit.
Specifications
  • Description:
    Quick-Test Auto-Scanning Tweezers, Straight Tips