Orders LinkContactUsLinkComponent
Super Thin and Long Tweezers, Electron Microscopy Sciences
Super Thin and Long Tweezers, Electron Microscopy Sciences
Super Thin and Long Tweezers, Electron Microscopy Sciences
Certificates
Super Thin and Long Tweezers, Electron Microscopy Sciences
Super Thin and Long Tweezers, Electron Microscopy Sciences
6 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These long and slender tweezers work well in multiple precision applications and are especially well suited for working near heat sources.
  • Made from SA steel
  • Sharp, relieve tips
  • Double bent tips allow more visibility
Product Family Options
Product Information
  • Length
    Tip style
    Availability
    Price
  • 100494-800
    Style 65A
    Style 65A
    140 mm
    Sharp, bent tips
    Log in for availability
    Log in for availability
    Each
    Each - $52.98
    $52.98
    Each
       $52.98
    Supplier Number: 78351-65A
  • 100493-878
    Style Stainless Steel
    Style Stainless Steel
    140 mm
    Long and narrow straight tips
    Log in for availability
    Log in for availability
    Each
    Each - $57.73
    $57.73
    Each
       $57.73
    Supplier Number: 72919-SS
  • 100494-802
    Style 66
    Style 66
    150 mm
    Sharp, double bent tips allow more visibility
    Log in for availability
    Log in for availability
    Each
    Each - $54.25
    $54.25
    Each
       $54.25
    Supplier Number: 78351-66
  • 100494-798
    Style 64A
    Style 64A
    140 mm
    Sharp, curve tips
    Log in for availability
    Log in for availability
    Each
    Each - $50.43
    $50.43
    Each
       $50.43
    Supplier Number: 78351-64A
  • 100494-804
    Style SSL
    Style SSL
    136 mm
    Long and narrow tips, ergonomic shape
    Log in for availability
    Log in for availability
    Each
    Each - $48.48
    $48.48
    Each
       $48.48
    Supplier Number: 78353-SSL
  • 100494-792
    Style 60
    Style 60
    140 mm
    Sharp, relieve tips
    Log in for availability
    Log in for availability
    Each
    Each - $68.04
    $68.04
    Each
       $68.04
    Supplier Number: 78348-60