About this item
Human unprocessed cord blood is collected aseptically at the time of delivery from healthy pregnant women
Cord blood is known to be a rich source of hematopoietic stem cells, which can be used to research hematopoietic and genetic disorders.
Serological and Infectious disease testing are performed with each donation. Customers should handle product as if potentially infectious following biosafety level 2 procedures.
The donor is required to review and sign an IRB approved procedure-specific consent form prior to the collection.