ContactUsLinkComponent
SP Bel-Art Tefzel® Forceps, Bel-Art Products, a part of SP
SP Bel-Art Tefzel® Forceps, Bel-Art Products, a part of SP
  16126-656
undefined
SP Bel-Art Tefzel® Forceps, Bel-Art Products, a part of SP
  16126-656
 :  H37922-0000
  • Length:
    11.4 cm (4½")
  • Cat. No.:
    16126-656
  • Supplier no.:
    379220000
Tefzel® Fluorocarbon plastic forceps have smooth square tips 1.6 mm (¹/₁₆") wide.
  • Fluorocarbon plastic construction
  • Smooth square tips
  • Autoclavable
They will withstand temperature up to 199ºC (390ºF) and are autoclavable. Excellent for handling small balance weights, they are individually blister packed.