ContactUsLinkComponent
RS-P26 Printer, METTLER TOLEDO®
RS-P26 Printer, METTLER TOLEDO®
Catalog # 97034-192
Supplier:  Mettler Toledo
undefined
RS-P26 Printer, METTLER TOLEDO®
Catalog # 97034-192
Supplier:  Mettler Toledo
Supplier Number:  11124323
Specifications
  • Description:
    RS-P26 printer
  • Cat. No.:
    97034-192
  • Electrical:
    100/240 V, 50/60 Hz
  • Print speed:
    2.3 lines/s
  • Supplier no.:
    11124323
  • Paper type:
    Standard paper
Specifications
About this item
The RS-P26 Compact Printer generates GxP-compliant printouts on regular or adhesive paper.
  • Compact design
  • Ergonomic
  • Easy operation
  • Allows taring and printing of balance
The ergonomic design makes operation easy and allows taring and printing of the balance directly from the printer.
The large paper rolls allow printing with fewer refills. The Printer automatically detects the settings of the connected device. Thanks to this feature no manual setting changes are required. The build-in real time clock and calendar combined with the lightfast paper ensure results documentation compliant with GxP. The Compact Printer supports the connected instrument languages, whether it operates in English, Russian, Chinese, Japanese or in any other language.
Thanks to the compact footprint, the RS-P26 printer finds place even on smallest work places.